Assay Details
| Gene Symbol: Kifc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010630.2 | NP_034760.2 | 9 - 10 | 1030 | 75 | |
| GenBank mRNA | AK144304.1 | - | 10 - 11 | 1189 | 75 | |
More Information
Set Membership: |
|

Kifc2
Genomic Map