Assay Details
| Gene Symbol: Cpsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164173.1 | NP_001157645.1 | 12 - 13 | 1412 | 109 | |
| NM_053193.2 | NP_444423.1 | 12 - 13 | 1412 | 109 | ||
| GenBank mRNA | AF322193.1 | - | 12 - 13 | 1383 | 109 | |
| AK149914.1 | - | 5 - 6 | 412 | 109 | ||
| AK155648.1 | - | 12 - 13 | 1353 | 109 | ||
| AK155806.1 | - | 12 - 13 | 1334 | 109 | ||
| BC011458.1 | - | 8 - 9 | 851 | 109 | ||
| BC049941.1 | - | 12 - 13 | 1234 | 109 | ||
| BC056388.1 | - | 12 - 13 | 1412 | 6854851 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf1
Genomic Map