Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 12 - 13 | 2001 | 86 | |
| GenBank mRNA | BC059932.1 | - | 10 - 11 | 2787 | 86 | |
| BC151196.1 | - | 7 - 8 | 1010 | 9088084 | 86 | |
More Information
Set Membership: |
|

Fbrsl1
Genomic Map