Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 14 - 15 | 2098 | 61 | |
| GenBank mRNA | AK129388.1 | - | 3 - 4 | 136 | 61 | |
| BC059932.1 | - | 12 - 13 | 2884 | 61 | ||
| BC069994.1 | - | 2 - 3 | 99 | 61 | ||
| BC151196.1 | - | 9 - 10 | 1107 | 9088084 | 61 | |
More Information
Set Membership: |
|

Fbrsl1
Genomic Map