Assay Details
| Gene Symbol: Ankle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253814.1 | NP_001240743.1 | 2 - 3 | 893 | 120 | |
| NM_027922.2 | NP_082198.1 | 2 - 3 | 893 | 120 | ||
| GenBank mRNA | AK043925.1 | - | 2 - 3 | 734 | 120 | |
| AK129192.1 | - | 1 - 2 | 219 | 120 | ||
| AK132187.1 | - | 2 - 3 | 895 | 120 | ||
| BC065071.1 | - | 2 - 3 | 856 | 6410033 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankle2
Genomic Map