Assay Details
| Gene Symbol: Ankle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253814.1 | NP_001240743.1 | 5 - 6 | 1490 | 105 | |
| NM_027922.2 | NP_082198.1 | 5 - 6 | 1487 | 105 | ||
| GenBank mRNA | AK003234.1 | - | 2 - 3 | 289 | 105 | |
| AK129192.1 | - | 4 - 5 | 816 | 105 | ||
| AK132187.1 | - | 5 - 6 | 1492 | 105 | ||
| AK142497.1 | - | 3 - 4 | 539 | 105 | ||
| BC065071.1 | - | 5 - 6 | 1450 | 6410033 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankle2
Genomic Map