Assay Details
| Gene Symbol: Ankle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253814.1 | NP_001240743.1 | 9 - 10 | 1954 | 60 | |
| NM_027922.2 | NP_082198.1 | 9 - 10 | 1951 | 60 | ||
| GenBank mRNA | AK003234.1 | - | 6 - 7 | 753 | 60 | |
| AK129192.1 | - | 8 - 9 | 1280 | 60 | ||
| AK132187.1 | - | 9 - 10 | 1956 | 60 | ||
| AK142497.1 | - | 7 - 8 | 1003 | 60 | ||
| BC065071.1 | - | 9 - 10 | 1914 | 6410033 | 60 | |
More Information
Set Membership: |
|

Ankle2
Genomic Map