Assay Details
| Gene Symbol: Plcxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281812.1 | NP_001268741.1 | 5 - 6 | 573 | 74 | |
| NM_207279.2 | NP_997162.2 | 6 - 7 | 738 | 74 | ||
| GenBank mRNA | AK039459.1 | - | 4 - 5 | 478 | 74 | |
| AK149435.1 | - | 5 - 6 | 574 | 74 | ||
| BC039627.1 | - | 6 - 7 | 738 | 5038680 | 74 | |
More Information
Set Membership: |
|

Plcxd1
Genomic Map