Assay Details
| Gene Symbol: Siglece | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031181.2 | NP_112458.2 | 3 - 4 | 797 | 82 | |
| GenBank mRNA | AF329269.1 | - | 3 - 4 | 801 | 82 | |
| AY348552.1 | - | 3 - 4 | 806 | 82 | ||
| AY371487.1 | - | 3 - 4 | 786 | 82 | ||
| BC023280.1 | - | 3 - 4 | 796 | 3662856 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglece
Genomic Map