Assay Details
| Gene Symbol: Ceacam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028236.1 | NP_082512.1 | 4 - 5 | 981 | 78 | |
| GenBank mRNA | AK008389.1 | - | 4 - 5 | 1013 | 78 | |
| BC042780.1 | - | 4 - 5 | 981 | 4224563 | 78 | |
More Information
Set Membership: |
|

Ceacam18
Genomic Map