Assay Details
| Gene Symbol: Ceacam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028236.1 | NP_082512.1 | 5 - 6 | 1118 | 64 | |
| GenBank mRNA | AK008389.1 | - | 5 - 6 | 1150 | 64 | |
| BC042780.1 | - | 5 - 6 | 1118 | 4224563 | 64 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370008 | AK008389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370008 | BC042780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370008 | NM_028236.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370008 | AK008389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370008 | BC042780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P370008 | NM_028236.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70927 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100565 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173453 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370008 | AK008389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370008 | BC042780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370008 | NM_028236.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370008 | AK008389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370008 | BC042780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P370008 | NM_028236.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083372 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100565 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01505052 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173453 | - | - |
| Whole Mouse Genome G4122A | A_51_P370008 | AK008389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370008 | BC042780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370008 | NM_028236.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370008 | AK008389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370008 | BC042780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370008 | NM_028236.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100565 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173453 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06246967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06246968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06382676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06479549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06479542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06063345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06410238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06410234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06210474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06008454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06623947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552457 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430927_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552451 | - | - |
| Mouse Genome 430 2.0 Array | 1430927_at | - | - |
| Mouse Genome 430 2.0 Array | 1430927_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72431_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72431_at | - | - |
| Mouse Expression Set 430 | 1430927_at | - | - |
| Mouse Expression Set 430 | 1430927_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430927_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430927_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 446075 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217029 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217029 | - | - |

Ceacam18
Genomic Map