Assay Details
| Gene Symbol: Cryba4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021351.1 | NP_067326.1 | 3 - 4 | 324 | 150 | |
| GenBank mRNA | AJ272228.2 | - | 3 - 4 | 324 | 150 | |
| AK132591.1 | - | 4 - 5 | 341 | 150 | ||
| AK162018.1 | - | 4 - 5 | 338 | 150 | ||
| BC056444.1 | - | 3 - 4 | 314 | 6824277 | 150 | |
| BC058703.1 | - | 3 - 4 | 324 | 6824293 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cryba4
Genomic Map