Assay Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163565.1 | NP_001157037.1 | 6 - 7 | 1028 | 80 | |
| NM_013643.2 | NP_038671.2 | 6 - 7 | 1136 | 80 | ||
| GenBank mRNA | AK038416.1 | - | 6 - 7 | 805 | 80 | |
| BC051975.1 | - | 6 - 7 | 777 | 80 | ||
| BC057339.1 | - | 5 - 6 | 1298 | 80 | ||
| BC079592.1 | - | 6 - 7 | 1108 | 6817360 | 80 | |
| S80329.1 | - | 3 - 4 | 972 | 80 | ||
| U28216.1 | - | 6 - 7 | 889 | 80 | ||
| U28217.1 | - | 6 - 7 | 1147 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn5
Genomic Map