Assay Details
| Gene Symbol: Cnot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271543.1 | NP_001258472.1 | 1 - 2 | 213 | 112 | |
| NM_011135.5 | NP_035265.1 | 1 - 2 | 213 | 112 | ||
| GenBank mRNA | AK007767.1 | - | 1 - 2 | 187 | 112 | |
| AK009561.1 | - | 1 - 2 | 201 | 112 | ||
| AK132063.1 | - | 1 - 2 | 133 | 112 | ||
| AK137838.1 | - | 1 - 2 | 178 | 112 | ||
| AK147048.1 | - | 1 - 2 | 180 | 112 | ||
| AK166449.1 | - | 1 - 2 | 177 | 112 | ||
| BC006021.1 | - | 1 - 2 | 157 | 3590764 | 112 | |
| CJ166755.1 | - | 1 - 2 | 213 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007767.1, AK132063.1, AK147048.1, AK166449.1, BC006021.1, NM_001271543.1, NM_011135.5 transcripts.
Set Membership: |
|

Cnot7
Genomic Map