Assay Details
| Gene Symbol: Cnot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271542.1 | NP_001258471.1 | 4 - 5 | 713 | 103 | |
| NM_001271543.1 | NP_001258472.1 | 4 - 5 | 784 | 103 | ||
| NM_011135.5 | NP_035265.1 | 4 - 5 | 784 | 103 | ||
| GenBank mRNA | AK044535.1 | - | 3 - 4 | 1650 | 103 | |
| AK132063.1 | - | 4 - 5 | 704 | 103 | ||
| AK135669.1 | - | 4 - 5 | 716 | 104 | ||
| AK147048.1 | - | 4 - 5 | 751 | 103 | ||
| AK166449.1 | - | 4 - 5 | 748 | 103 | ||
| BC006021.1 | - | 4 - 5 | 728 | 3590764 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot7
Genomic Map