Assay Details
| Gene Symbol: Zdhhc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178395.3 | NP_848482.1 | 4 - 5 | 603 | 77 | |
| GenBank mRNA | AK018284.1 | - | 575 | 77 | ||
| AK030662.1 | - | 4 - 5 | 604 | 77 | ||
| AK044652.1 | - | 4 - 5 | 597 | 77 | ||
| AK136296.1 | - | 4 - 5 | 602 | 77 | ||
| BC117761.1 | - | 5 - 6 | 365 | 77 | ||
| BC127156.1 | - | 5 - 6 | 437 | 40090859 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc2
Genomic Map