Assay Details
| Gene Symbol: Zdhhc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178395.3 | NP_848482.1 | 5 - 6 | 662 | 74 | |
| GenBank mRNA | AK018284.1 | - | 634 | 74 | ||
| AK030662.1 | - | 5 - 6 | 663 | 74 | ||
| AK044652.1 | - | 5 - 6 | 656 | 74 | ||
| AK136296.1 | - | 5 - 6 | 661 | 74 | ||
| BC117761.1 | - | 6 - 7 | 424 | 74 | ||
| BC127156.1 | - | 6 - 7 | 496 | 40090859 | 74 | |
| Z47777.1 | - | 2 - 3 | 74 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc2
Genomic Map