Assay Details
| Gene Symbol: Vav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020505.2 | NP_065251.2 | 25 - 26 | 2711 | 77 | |
| NM_146139.2 | NP_666251.1 | 8 - 9 | 749 | 77 | ||
| GenBank mRNA | AK147053.1 | - | 8 - 9 | 726 | 77 | |
| BC027242.1 | - | 8 - 9 | 697 | 77 | ||
| BC052739.1 | - | 25 - 26 | 2458 | 6832850 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vav3
Genomic Map