Assay Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190483.1 | NP_001177412.1 | 37 - 38 | 5729 | 73 | |
| GenBank mRNA | D17583.1 | - | 30 - 31 | 4262 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcsk5
Genomic Map