Assay Details
| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152923.2 | NP_690887.2 | 10 - 11 | 1470 | 71 | |
| GenBank mRNA | AY118171.1 | - | 10 - 11 | 1470 | 71 | |
| BC116306.1 | - | 10 - 11 | 1461 | 40095195 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnq3
Genomic Map