Assay Details
| Gene Symbol: Efr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133766.3 | NP_598527.2 | 16 - 17 | 2063 | 101 | |
| GenBank mRNA | AB158474.1 | - | 16 - 17 | 2067 | 101 | |
| AK018695.1 | - | 11 - 12 | 1278 | 101 | ||
| AK030074.1 | - | 16 - 17 | 2064 | 101 | ||
| AK032461.1 | - | 16 - 17 | 2036 | 101 | ||
| AK044000.1 | - | 16 - 17 | 2057 | 101 | ||
| AK044475.1 | - | 16 - 17 | 2059 | 101 | ||
| AK129065.1 | - | 16 - 17 | 2041 | 101 | ||
| AK161250.1 | - | 16 - 17 | 2048 | 101 | ||
| BC007482.1 | - | 16 - 17 | 2067 | 3495608 | 101 | |
| BC032092.1 | - | 16 - 17 | 2068 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efr3a
Genomic Map