Assay Details
| Gene Symbol: Efr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133766.3 | NP_598527.2 | 8 - 9 | 1056 | 113 | |
| GenBank mRNA | AB158474.1 | - | 8 - 9 | 1060 | 113 | |
| AK018695.1 | - | 3 - 4 | 276 | 113 | ||
| AK030074.1 | - | 8 - 9 | 1057 | 113 | ||
| AK032461.1 | - | 8 - 9 | 1029 | 113 | ||
| AK044000.1 | - | 8 - 9 | 1050 | 113 | ||
| AK044475.1 | - | 8 - 9 | 1052 | 113 | ||
| AK129065.1 | - | 8 - 9 | 1034 | 113 | ||
| AK161250.1 | - | 8 - 9 | 1041 | 113 | ||
| BC007482.1 | - | 8 - 9 | 1060 | 3495608 | 113 | |
| BC032092.1 | - | 8 - 9 | 1061 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efr3a
Genomic Map