Assay Details
| Gene Symbol: Oc90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010953.2 | NP_035083.1 | 10 - 11 | 814 | 76 | |
| GenBank mRNA | AF093591.1 | - | 10 - 11 | 924 | 76 | |
| AK012981.1 | - | 10 - 11 | 816 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oc90
Genomic Map