Assay Details
| Gene Symbol: Oc90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010953.2 | NP_035083.1 | 9 - 10 | 773 | 63 | |
| GenBank mRNA | AK012981.1 | - | 9 - 10 | 775 | 63 | |
More Information
Set Membership: |
|

Oc90
Genomic Map