Assay Details
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198019.2 | NP_932136.2 | 12 - 13 | 1581 | 85 | |
GenBank mRNA | AK050870.1 | - | 11 - 12 | 1707 | 85 | |
BC070400.1 | - | 12 - 13 | 1581 | 5694341 | 85 | |
BC071270.1 | - | 12 - 13 | 1563 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
