Assay Details
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198019.2 | NP_932136.2 | 5 - 6 | 909 | 86 | |
GenBank mRNA | AK050870.1 | - | 5 - 6 | 616 | 86 | |
BC070400.1 | - | 5 - 6 | 909 | 5694341 | 86 | |
BC071270.1 | - | 5 - 6 | 892 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
