Assay Details
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198019.2 | NP_932136.2 | 10 - 11 | 1374 | 101 | |
GenBank mRNA | AK050870.1 | - | 9 - 10 | 1500 | 101 | |
AK160295.1 | - | 3 - 4 | 605 | 101 | ||
BC070400.1 | - | 10 - 11 | 1374 | 5694341 | 101 | |
BC071270.1 | - | 10 - 11 | 1356 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
