Assay Details
| Gene Symbol: Eif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133916.2 | NP_598677.1 | 11 - 12 | 1701 | 65 | |
| GenBank mRNA | AK033722.1 | - | 11 - 12 | 1701 | 65 | |
| AK089978.1 | - | 11 - 12 | 1690 | 65 | ||
| AK167618.1 | - | 11 - 12 | 1688 | 65 | ||
| AK170938.1 | - | 11 - 12 | 1701 | 65 | ||
| BC007175.1 | - | 10 - 11 | 1160 | 65 | ||
| BC023767.1 | - | 11 - 12 | 1672 | 5343631 | 65 | |
| BC031704.1 | - | 11 - 12 | 1678 | 4023277 | 65 | |
| BC051065.1 | - | 11 - 12 | 1661 | 5343631 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3b
Genomic Map