Assay Details
| Gene Symbol: Eif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133916.2 | NP_598677.1 | 1 - 2 | 527 | 74 | |
| GenBank mRNA | AK033722.1 | - | 1 - 2 | 527 | 74 | |
| AK089978.1 | - | 1 - 2 | 515 | 74 | ||
| AK167618.1 | - | 1 - 2 | 514 | 74 | ||
| AK170938.1 | - | 1 - 2 | 527 | 74 | ||
| BC023767.1 | - | 1 - 2 | 498 | 5343631 | 74 | |
| BC031704.1 | - | 1 - 2 | 504 | 4023277 | 74 | |
| BC051065.1 | - | 1 - 2 | 487 | 5343631 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3b
Genomic Map