Assay Details
| Gene Symbol: Eif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133916.2 | NP_598677.1 | 8 - 9 | 1378 | 108 | |
| GenBank mRNA | AK089978.1 | - | 8 - 9 | 1366 | 108 | |
| AK167618.1 | - | 8 - 9 | 1365 | 108 | ||
| AK170938.1 | - | 8 - 9 | 1378 | 108 | ||
| BC007175.1 | - | 7 - 8 | 837 | 108 | ||
| BC023767.1 | - | 8 - 9 | 1349 | 5343631 | 108 | |
| BC031704.1 | - | 8 - 9 | 1355 | 4023277 | 108 | |
| BC051065.1 | - | 8 - 9 | 1338 | 5343631 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3b
Genomic Map