Assay Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133349.3 | NP_579927.1 | 1 - 2 | 221 | 96 | |
| GenBank mRNA | AK146082.1 | - | 1 - 2 | 159 | 96 | |
| AK171245.1 | - | 1 - 2 | 161 | 96 | ||
| BC058780.1 | - | 1 - 2 | 147 | 6312202 | 96 | |
| CJ083782.1 | - | 1 - 2 | 277 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146082.1, AK171245.1, BC058780.1, NM_133349.3 transcripts.
Set Membership: |
|

Zfand2a
Genomic Map