Assay Details
| Gene Symbol: Marveld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183195.2 | NP_899018.1 | 1 - 2 | 2167 | 87 | |
| GenBank mRNA | AK132243.1 | - | 1 - 2 | 2158 | 87 | |
| BC044794.1 | - | 1 - 2 | 593 | 87 | ||
| BC054384.1 | - | 1 - 2 | 2130 | 6475446 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119296 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119293 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158814 | AK081212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P158814 | AK081212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | BC054384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | AK132243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | BC044794.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | NM_183195.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | BC054384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | AK132243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | BC044794.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541175 | NM_183195.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157104 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | BC054384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | AK132243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | BC044794.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | NM_183195.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | BC054384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | AK132243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | BC044794.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541175 | NM_183195.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158814 | AK081212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P158814 | AK081212.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32119356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157104 | - | - |
| Whole Mouse Genome G4122A | A_52_P541175 | BC054384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | AK132243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | BC044794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | NM_183195.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | BC054384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | AK132243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | BC044794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541175 | NM_183195.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158814 | AK081212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158814 | AK081212.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157104 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119293 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119296 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32119292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14327718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14350742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14319415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14319418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14151364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14393904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32119282 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10463226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463225 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436830_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436773_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439381_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433538_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10463224 | - | - |
| Mouse Genome 430 2.0 Array | 1436830_at | - | - |
| Mouse Genome 430 2.0 Array | 1433538_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439381_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436773_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1433538_at | - | - |
| Mouse Genome 430 2.0 Array | 1436773_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439381_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436830_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277010_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277010_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900005457.mm.1 | NM_183195.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005457.mm.1 | NM_183195.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005457.mm.1 | TC1900000610.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900005458.mm.1 | NM_183195.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005458.mm.1 | NM_183195.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005458.mm.1 | TC1900000610.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900005456.mm.1 | NM_183195.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005456.mm.1 | NM_183195.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005456.mm.1 | TC1900000610.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900005460.mm.1 | NM_183195.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005460.mm.1 | NM_183195.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005460.mm.1 | TC1900000610.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900005461.mm.1 | NM_183195.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005461.mm.1 | NM_183195.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900005461.mm.1 | TC1900000610.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433538_PM_at | - | - |
| Mouse Expression Set 430 | 1433538_at | - | - |
| Mouse Expression Set 430 | 1436773_PM_x_at | - | - |
| Mouse Expression Set 430 | 1439381_PM_x_at | - | - |
| Mouse Expression Set 430 | 1436773_x_at | - | - |
| Mouse Expression Set 430 | 1439381_x_at | - | - |
| Mouse Expression Set 430 | 1436830_at | - | - |
| Mouse Expression Set 430 | 1436830_PM_at | - | - |
| Murine Genome U74v2 Set | 98036_at | - | - |
| Murine Genome U74v2 Set | 98036_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436773_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433538_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439381_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436773_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433538_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439381_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433538_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439381_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436773_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439381_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436773_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433538_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436830_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436830_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 747826 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 107100095 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107100095 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238644 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238644 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259554 | - | - |
| mouse-6 v1.1 (Illumina) | scl53115.2.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107100095 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53115.2.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107100095 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110309 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238644 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259554 | - | - |

Marveld1
Genomic Map