Assay Details
| Gene Symbol: Srrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109909.1 | NP_001103379.1 | 5 - 6 | 855 | 85 | |
| NM_001109910.1 | NP_001103380.1 | 5 - 6 | 855 | 85 | ||
| NM_031405.2 | NP_113582.1 | 5 - 6 | 855 | 85 | ||
| GenBank mRNA | AK144300.1 | - | 5 - 6 | 846 | 85 | |
| BC066831.1 | - | 5 - 6 | 814 | 30471339 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srrt
Genomic Map