Assay Details
| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031406.3 | NP_113583.2 | 1 - 2 | 131 | 70 | |
| GenBank mRNA | AK050658.1 | - | 1 - 2 | 146 | 70 | |
| AK154118.1 | - | 1 - 2 | 89 | 70 | ||
| BC046982.1 | - | 1 - 2 | 51 | 6478046 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC046982.1, NM_031406.3 transcripts.
Set Membership: |
|

Slc12a9
Genomic Map