Assay Details
| Gene Symbol: Muc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF027131.1 | - | 4 - 5 | 559 | 158 | |
| BC015298.1 | - | 3 - 4 | 405 | 158 | ||
| BC026665.1 | - | 3 - 4 | 241 | 158 | ||
| BC046639.1 | - | 2 - 3 | 146 | 158 | ||
More Information
Set Membership: |
|

Muc3
Genomic Map