Assay Details
| Gene Symbol: Muc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF027131.1 | - | 6 - 7 | 789 | 59 | |
| BC015298.1 | - | 5 - 6 | 635 | 59 | ||
| BC026665.1 | - | 5 - 6 | 471 | 59 | ||
| BC046639.1 | - | 4 - 5 | 376 | 59 | ||
| BC058768.1 | - | 4 - 5 | 318 | 59 | ||
More Information
Set Membership: |
|

Muc3
Genomic Map