Assay Details
| Gene Symbol: Gm20605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033148.1 | 7 - 8 | 1032 | 71 | ||
| Gene Symbol: Lrch4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168652.1 | NP_001162123.1 | 7 - 8 | 1032 | 71 | |
| NM_146164.2 | NP_666276.1 | 7 - 8 | 1032 | 71 | ||
| GenBank mRNA | AK088331.1 | - | 7 - 8 | 978 | 71 | |
| BC012525.1 | - | 7 - 8 | 972 | 2598844 | 71 | |
| CF729493.1 | - | 5 - 6 | 547 | 71 | ||
| DV661712.1 | - | 4 - 5 | 459 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm20605
Genomic Map