Assay Details
| Gene Symbol: Gm20605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033148.1 | 8 - 9 | 1120 | 96 | ||
| Gene Symbol: Lrch4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168652.1 | NP_001162123.1 | 8 - 9 | 1120 | 96 | |
| NM_146164.2 | NP_666276.1 | 8 - 9 | 1120 | 96 | ||
| GenBank mRNA | AK088331.1 | - | 8 - 9 | 1066 | 96 | |
| AK131167.1 | - | 1280 | 96 | |||
| BC012525.1 | - | 8 - 9 | 1060 | 2598844 | 96 | |
| CF729493.1 | - | 6 - 7 | 635 | 96 | ||
| DV661712.1 | - | 5 - 6 | 547 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm20605
Genomic Map