Assay Details
| Gene Symbol: Fam102b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163567.1 | NP_001157039.1 | 10 - 11 | 1354 | 65 | |
| GenBank mRNA | AK046595.1 | - | 10 - 11 | 1020 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196629 | AK046595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196629 | NM_001163567.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196629 | AK046595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196629 | NM_001163567.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591801 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482172 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4329 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64200 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117585 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129308 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196629 | AK046595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196629 | NM_001163567.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196629 | AK046595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196629 | NM_001163567.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05026692 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05026601 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117585 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05026576 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129308 | - | - |
| Whole Mouse Genome G4122A | A_51_P502724 | AK005412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P502724 | NM_001163567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196629 | AK046595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196629 | NM_001163567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196629 | AK046595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196629 | NM_001163567.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129308 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02512529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02663389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02601919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02601923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02950908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03017349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02344202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02526973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03078911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03124101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02859717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03124100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02501470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02389270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02937981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02931693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03017343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02810155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02661008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02389271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02323150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02911245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02865323 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1434828_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455033_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501456 | - | - |
| Mouse Genome 430 2.0 Array | 1434828_at | - | - |
| Mouse Genome 430 2.0 Array | 1455033_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455033_at | - | - |
| Mouse Genome 430 2.0 Array | 1434828_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329739_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329739_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300021744.mm.1 | NM_001163567.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021744.mm.1 | NM_001163567.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021744.mm.1 | TC0300002760.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455033_at | - | - |
| Mouse Expression Set 430 | 1434828_PM_at | - | - |
| Mouse Expression Set 430 | 1455033_PM_at | - | - |
| Mouse Expression Set 430 | 1434828_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455033_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434828_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434828_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455033_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253691 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253691 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247740 | - | - |

Fam102b
Genomic Map