Assay Details
| Gene Symbol: Fam102b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163567.1 | NP_001157039.1 | 5 - 6 | 737 | 140 | |
| GenBank mRNA | AK046595.1 | - | 5 - 6 | 403 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam102b
Genomic Map