Assay Details
| Gene Symbol: Glt1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177005.4 | NP_795979.2 | 4 - 5 | 399 | 79 | |
| GenBank mRNA | AK077577.1 | - | 4 - 5 | 401 | 79 | |
| BC115464.1 | - | 4 - 5 | 398 | 79 | ||
More Information
Set Membership: |
|

Glt1d1
Genomic Map