Assay Details
| Gene Symbol: Tlr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126166.4 | NP_569054.2 | 3 - 4 | 351 | 139 | |
| GenBank mRNA | AK083977.1 | - | 3 - 4 | 352 | 139 | |
| AK087739.1 | - | 2 - 3 | 173 | 139 | ||
| AK166177.1 | - | 2 - 3 | 226 | 139 | ||
| AK171438.1 | - | 3 - 4 | 334 | 139 | ||
| BC099937.1 | - | 3 - 4 | 389 | 30084049 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083977.1, AK087739.1, AK166177.1, BC099937.1, NM_126166.4 transcripts.
Set Membership: |
|

Tlr3
Genomic Map