Assay Details
| Gene Symbol: Olfr1449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146303.1 | NP_666415.1 | 1 | 455 | 76 | |
| GenBank mRNA | BC133705.1 | - | 1 | 459 | 40135405 | 76 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002978 | NM_146303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2002978 | BC133705.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180784 | NM_146303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180784 | BC133705.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75314 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164025 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180784 | NM_146303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180784 | BC133705.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002978 | NM_146303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2002978 | BC133705.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P494257 | NM_146303.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P494257 | BC133705.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963111 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164025 | - | - |
| Whole Mouse Genome G4122A | A_51_P494257 | NM_146303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P494257 | BC133705.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130633 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164025 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436066 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436070 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14436072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14223582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14477670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14494120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461785 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461784 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900003023.mm.1 | NM_146303.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003023.mm.1 | NM_146303.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003023.mm.1 | TC1900000260.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 670195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223962 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223962 | - | - |
| mouse-6 v1.1 (Illumina) | scl6293.1.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl6293.1.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223962 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223962 | - | - |

Olfr1449
Genomic Map