Assay Details
| Gene Symbol: Uhrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144873.2 | NP_659122.2 | 4 - 5 | 1131 | 106 | |
| GenBank mRNA | AB116653.1 | - | 4 - 5 | 866 | 106 | |
| AK080925.1 | - | 4 - 5 | 1172 | 106 | ||
| AK082518.1 | - | 3 - 4 | 942 | 106 | ||
| BC060241.1 | - | 4 - 5 | 1131 | 6334856 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uhrf2
Genomic Map