Assay Details
| Gene Symbol: Uhrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144873.2 | NP_659122.2 | 7 - 8 | 1551 | 115 | |
| GenBank mRNA | AB116653.1 | - | 7 - 8 | 1286 | 115 | |
| AF274047.1 | - | 7 - 8 | 1461 | 115 | ||
| AK080925.1 | - | 7 - 8 | 1592 | 115 | ||
| BC060241.1 | - | 7 - 8 | 1551 | 6334856 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uhrf2
Genomic Map