Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 13 - 14 | 1819 | 72 | |
| GenBank mRNA | AK082884.1 | - | 13 - 14 | 1810 | 72 | |
| BC027065.1 | - | 8 - 9 | 893 | 72 | ||
| BC039634.1 | - | 2 - 3 | 243 | 72 | ||
| BC068126.1 | - | 13 - 14 | 1727 | 6829456 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyw1
Genomic Map