Assay Details
| Gene Symbol: Tyw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015876.2 | NP_001015876.1 | 9 - 10 | 1277 | 79 | |
| GenBank mRNA | AK082884.1 | - | 9 - 10 | 1268 | 79 | |
| BC027065.1 | - | 4 - 5 | 351 | 79 | ||
| BC068126.1 | - | 9 - 10 | 1185 | 6829456 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tyw1
Genomic Map