Assay Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145218.3 | NP_660253.2 | 4 - 5 | 1683 | 84 | |
| GenBank mRNA | AB175684.1 | - | 4 - 5 | 761 | 84 | |
| AF467979.1 | - | 4 - 5 | 964 | 84 | ||
| AK035817.1 | - | 4 - 5 | 1437 | 84 | ||
| AK051281.1 | - | 4 - 5 | 2083 | 84 | ||
| AK087726.1 | - | 2253 | 84 | |||
| BC052469.1 | - | 4 - 5 | 1079 | 6810287 | 84 | |
| BC069177.1 | - | 4 - 5 | 1683 | 84 | ||
| BC158110.1 | - | 4 - 5 | 1710 | 9088184 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wbscr17
Genomic Map