Assay Details
| Gene Symbol: Gk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010294.1 | NP_034424.1 | 1 | 1717 | 139 | |
| GenBank mRNA | AF117734.1 | - | 1 | 1717 | 139 | |
| AK075761.1 | - | 1 | 1630 | 139 | ||
| BC050764.1 | - | 1 | 1730 | 6774427 | 139 | |
| BC061147.1 | - | 1 | 1696 | 6770827 | 139 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
glycerol metabolic process
glycerol-3-phosphate metabolic process
triglyceride metabolic process
phosphorylation
glycerol-3-phosphate biosynthetic process
nucleotide binding
glycerol kinase activity
ATP binding
kinase activity
transferase activity
phosphotransferase activity, alcohol group as acceptor
carbohydrate metabolic process
glycerol metabolic process
glycerol-3-phosphate metabolic process
triglyceride metabolic process
phosphorylation
glycerol-3-phosphate biosynthetic process
nucleotide binding
glycerol kinase activity
ATP binding
kinase activity
transferase activity
phosphotransferase activity, alcohol group as acceptor
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973235 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131647 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23912783 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131647 | - | - |
| Whole Mouse Genome G4122A | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | BC050764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | BC061147.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | NM_010294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | AF117734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P194658 | AK075761.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108207 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131647 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04931086 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04931087 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04803516 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04931112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04931097 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04931100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04803515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23912772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04502514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04931090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04425110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04425121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04931093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04390596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04435679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04390589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04615402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04390591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04931100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04794932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04862422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23912747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531559 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531558 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416694_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531556 | - | - |
| Mouse Genome 430 2.0 Array | 1416694_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416694_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14626_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14626_PM_at | - | - |
| Mouse Expression Set 430 | 1416694_PM_at | - | - |
| Mouse Expression Set 430 | 1416694_at | - | - |
| Murine Genome U74v2 Set | 165413_at | - | - |
| Murine Genome U74v2 Set | 165413_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416694_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416694_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416694_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416694_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 657481 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6510736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006510736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259416 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259416 | - | - |
| mouse-6 v1.1 (Illumina) | scl26349.2.101_165-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6510736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26349.2.101_165-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1259416 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259416 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl26349.2.101_165-S | - | - |

Gk2
Genomic Map