Assay Details
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 15 - 16 | 1775 | 91 | |
GenBank mRNA | AK145898.1 | - | 14 - 15 | 2262 | 91 | |
BC060184.1 | - | 14 - 15 | 2216 | 91 | ||
BC139010.1 | - | 15 - 16 | 1705 | 8862032 | 91 | |
BC139011.1 | - | 15 - 16 | 1705 | 8862033 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
